Order Entry
Denmark
ContactUsLinkComponent
Protective hand creams, Lindesa® K with camomile
Protective hand creams, Lindesa® K with camomile
Catalog # FAWE13642006
Supplier:  FAWECO HAUTSCHUTZ
undefined
Protective hand creams, Lindesa® K with camomile
Catalog # FAWE13642006
Supplier:  FAWECO HAUTSCHUTZ
Specifications
  • Description:
    Protective hand cream, Lindesa® K with camomile, 50 ml tube
  • Pk:
    24
Specifications
About this item
The cream strengthens the skin's immunity, prevents skin damage from exposure to external contamination, and soothes irritated skin without leaving a greasy film. It also is suitable for the conditioning of stressed skin.
  • Does not leave any grease marks and is quickly absorbed
  • O/w emulsion
  • Silicone-free, perfumed
Ideal for use in workplaces where no greasy residue can be left on surfaces, such as when varnishing, in the production of electronics, or during final inspection and testing.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904