Goggles, uvex ultrasonic
Supplier: UVEX
Certificates
About this item
These full view goggles fit individual facial features well, due to their combination of hard and soft materials, ensuring a high level of comfort for the wearer.
- Optimised internal volume and extensive side protection, even for spectacle wearers
- Prescription insert possible
- Wide panoramic field of view
- Ventilated for a pleasant and irritation-free interior environment
Weight: 80 g.
* Reduced top and bottom ventilation. ** Rubber headband.
** Model has rubber headband.
Certifications: EN 166, EN 170; 1B.
Frequently Bought Together