Order Entry
Denmark
Orders LinkContactUsLinkComponent
Büchner funnels, DURAN®
Supplier:   DWK Life Sciences
Büchner funnels, DURAN®
Supplier:  DWK Life Sciences
Certificates
Büchner funnels, DURAN®
Büchner funnels, DURAN®
5 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
For filter papers or membrane filtration discs.
Product Family Options
Product Information
  • Plate Ø
    Capacity
    For filter Ø
    Availability
    Price
  • 48 mm
    70 ml
    45 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr862.00
    kr862.00
    1 * 1 Items
       kr862.00
  • 60 mm
    125 ml
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr1,030.00
    kr1,030.00
    1 * 1 Items
       kr1,030.00
  • 73 mm
    220 ml
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr1,360.00
    kr1,360.00
    1 * 1 Items
       kr1,360.00
  • 95 mm
    500 ml
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr1,540.00
    kr1,540.00
    1 * 1 Items
       kr1,540.00
  • 120 mm
    1000 ml
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr2,160.00
    kr2,160.00
    1 * 1 Items
       kr2,160.00