Order Entry
Denmark
Orders LinkContactUsLinkComponent
Büchner funnels
Büchner funnels
Supplier:   KARTELL
Büchner funnels
Supplier:  KARTELL
Certificates
Büchner funnels
Büchner funnels
8 Options Available Below
Product Details & Documents
PP, white.
  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning
For filter paper or membrane filtration discs.
Product Family Options
Product Information
  • Capacity
    For filter Ø
    Availability
    Price
  • 40 ml
    42,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 168,00 kr
    168,00 kr
    1 * 1 Items
       168,00 kr
  • 70 ml
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 181,00 kr
    181,00 kr
    1 * 1 Items
       181,00 kr
  • 180 ml
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 174,00 kr
    174,00 kr
    1 * 1 Items
       174,00 kr
  • 235 ml
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 140,00 kr
    140,00 kr
    1 * 1 Items
       140,00 kr
  • 390 ml
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 285,00 kr
    285,00 kr
    1 * 1 Items
       285,00 kr
  • 810 ml
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 380,00 kr
    380,00 kr
    1 * 1 Items
       380,00 kr
  • 2100 ml
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 450,00 kr
    450,00 kr
    1 * 1 Items
       450,00 kr
  • 6000 ml
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 767,00 kr
    767,00 kr
    1 * 1 Items
       767,00 kr