Order Entry
Denmark
Orders LinkContactUsLinkComponent
Powder funnels, Quickfit®
Powder funnels, Quickfit®
Supplier:   SciLabware
Powder funnels, Quickfit®
Supplier:  SciLabware
Certificates
Powder funnels, Quickfit®
Powder funnels, Quickfit®
5 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, stem with cone.
  • Very good chemical resistance
  • High temperature resistance
Designed for transfer of powders into flasks and vessels.
Product Family Options
Product Information
  • Cone size
    Funnel O-Ø
    Availability
    Price
  • 14/23 NS
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 390,00 kr
    390,00 kr
    1 * 1 Items
       390,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    201-8294
  • 19/26 NS
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 390,00 kr
    390,00 kr
    1 * 1 Items
       390,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    201-8295
  • 24/29 NS
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 390,00 kr
    390,00 kr
    1 * 1 Items
       390,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    201-8296
  • 29/32 NS
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 457,00 kr
    457,00 kr
    1 * 1 Items
       457,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    532-0072
  • 34/35 NS
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 457,00 kr
    457,00 kr
    1 * 1 Items
       457,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    532-0073