Order Entry
Denmark
Orders LinkContactUsLinkComponent
Forceps
Forceps
105 mm
Catalog # 232-0039
Supplier:   MURRAY S
Forceps
Catalog # 232-0039
Supplier:  MURRAY S
Forceps
Forceps
105 mm
Price
80,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Steel, nickel plated
  • With flat, curved ends
  • Robust with easy action
  • Very good thermal resistance, good chemical resistance
  • Light and easy to clean
Ideal for cover glasses, slides and filter discs.
Specifications
  • Length:
    105 mm