Order Entry
Denmark
Orders LinkContactUsLinkComponent
Rotary evaporating flasks
Rotary evaporating flasks
Supplier:   SciLabware
Rotary evaporating flasks
Supplier:  SciLabware
Certificates
Rotary evaporating flasks
Rotary evaporating flasks
10 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for use with rotary evaporators. Large surface area for rapid evaporation. Robust heavy wall construction for added strength. Florentine shape allows easy removal of solid residues.
Product Family Options
Product Information
  • Capacity
    Socket size
    Total height
    Availability
    Price
  • 50 ml
    29/32 NS
    94 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 331,00 kr
    331,00 kr
    1 * 1 Items
       331,00 kr
  • 100 ml
    29/32 NS
    107 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 377,00 kr
    377,00 kr
    1 * 1 Items
       377,00 kr
  • 100 ml
    24/29 NS
    108 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 368,00 kr
    368,00 kr
    1 * 1 Items
       368,00 kr
  • 250 ml
    24/29 NS
    148 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 1.990,00 kr
    1.990,00 kr
    1 * 5 Items
       1.990,00 kr
  • 250 ml
    29/32 NS
    151 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 402,00 kr
    402,00 kr
    1 * 1 Items
       402,00 kr
  • 500 ml
    29/32 NS
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 595,00 kr
    595,00 kr
    1 * 1 Items
       595,00 kr
  • 500 ml
    24/29 NS
    165 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 2.830,00 kr
    2.830,00 kr
    1 * 5 Items
       2.830,00 kr
  • 1000 ml
    29/32 NS
    198 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 548,00 kr
    548,00 kr
    1 * 1 Items
       548,00 kr
  • 1000 ml
    24/29 NS
    204 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 3.280,00 kr
    3.280,00 kr
    1 * 5 Items
       3.280,00 kr
  • 2000 ml
    29/32 NS
    245 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 926,00 kr
    926,00 kr
    1 * 1 Items
       926,00 kr