Flasks, pear shaped with standard ground joint
Supplier: Lenz Laborglas
Jump to:
About this item
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint socket and white labelling area.
13 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint socket and white labelling area.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Product Family Options
Product Information
- CapacitySocket sizeHeightExt.ØAvailabilityPrice