Order Entry
Denmark
Orders LinkContactUsLinkComponent
Caps, ERLENMEYER-CAP
Caps, ERLENMEYER-CAP
Supplier:   LUDI
Caps, ERLENMEYER-CAP
Supplier:  LUDI
Certificates
Caps, ERLENMEYER-CAP
Caps, ERLENMEYER-CAP
3 Options Available Below
Product Details & Documents
Aluminium, anodised.
  • Springs made of chromium-nickel steel
  • Springs clamp against the outer wall of the vessel
  • Autoclavable
Product Family Options
Product Information
  • Colour
    For
    Availability
    Price
  • Silver
    37-39 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - kr610.00
    kr610.00
    1 * 10 Items
       kr610.00
  • Red
    37-39 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - kr610.00
    kr610.00
    1 * 10 Items
       kr610.00
  • Blue
    37-39 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - kr610.00
    kr610.00
    1 * 10 Items
       kr610.00