Capsule filters, Polycap™ TC, Whatman™
These Polycap™ TC capsule filters feature dual layer polytehersulfone (PES) membranes and provide efficient filtration for aqueous solutions.
- 100% integrity-tested during manufacturing
- PP housing thermally fused to eliminate surfactants and mold releasing agents that could affect analysis
- Available in sterile and non sterile versions with a filling bell option
- PES membranes protein adsorption characteristics: HAS 0,4 µg/cm²; Insulin 2,0 µg/cm²; gammaglobulin 1,5 µg/cm²
PES membranes are inherently hydrophilic, have low extractables, are biosafe, and have excellent flow rates. These characteristics make Polycap™ TC PES filters appropriate for a variety of critical filtration applications, including cleaning/rinsing pharmaceutical solutions, tissue culture media, and viral suspensions.
Polycap™ AS (Aqueous Solution) - recommended for filtering aqueous solutions. It combines a glass microfibre (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.
Polycap™ TF - designed for use with organic solvents and chemically aggressive solutions.
Polycap™ HD (Heavy Duty) - offers high filtration efficiency and excellent filtrate purity due to its materials and methods of manufacture.
Polycap™ SPF (Serum Prefilter) - optimised for prefiltration applications and typically used upstream of a Polycap™ AS or a Polycap™ PES capsule.
: Manufactured according to ISO9001:2008 QMS standards
These Polycap™ TC capsule filters feature dual layer polytehersulfone (PES) membranes and provide efficient filtration for aqueous solutions.
- 100% integrity-tested during manufacturing
- PP housing thermally fused to eliminate surfactants and mold releasing agents that could affect analysis
- Available in sterile and non sterile versions with a filling bell option
- PES membranes protein adsorption characteristics: HAS 0,4 µg/cm²; Insulin 2,0 µg/cm²; gammaglobulin 1,5 µg/cm²
PES membranes are inherently hydrophilic, have low extractables, are biosafe, and have excellent flow rates. These characteristics make Polycap™ TC PES filters appropriate for a variety of critical filtration applications, including cleaning/rinsing pharmaceutical solutions, tissue culture media, and viral suspensions.
Polycap™ AS (Aqueous Solution) - recommended for filtering aqueous solutions. It combines a glass microfibre (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.
Polycap™ TF - designed for use with organic solvents and chemically aggressive solutions.
Polycap™ HD (Heavy Duty) - offers high filtration efficiency and excellent filtrate purity due to its materials and methods of manufacture.
Polycap™ SPF (Serum Prefilter) - optimised for prefiltration applications and typically used upstream of a Polycap™ AS or a Polycap™ PES capsule.
: Manufactured according to ISO9001:2008 QMS standards
- Catalog No:
- 515-2531
- 515-2532
- 515-2533
- 515-2538
- 515-2536
- 515-2537
- 516-0983
- 515-2540
- 515-2541
- 515-2546
- 515-2547
- 515-2544
- 516-0984
- 515-2551
- 516-0915
- 515-2552
- 515-2553
- 515-2555
- 516-0985
- Type:
- Polycap™ TC 36, PES, sterile
- Polycap™ TC 36, PES, sterile
- Polycap™ TC 36, PES, sterile
- Polycap™ TC 36, PES, with filling bell, non sterile
- Polycap™ TC 36, PES, with filling bell, sterile
- Polycap™ TC 36, PES, with filling bell, sterile
- Polycap™ TC 36, PES, with filling bell, sterile
- Polycap™ TC 75, PES, sterile
- Polycap™ TC 75, PES, sterile
- Polycap™ TC 75, PES, sterile
- Polycap™ TC 75, PES, sterile
- Polycap™ TC 75, PES, with filling bell, sterile
- Polycap™ TC 75, PES, with filling bell, sterile
- Polycap™ TC 150, PES, sterile
- Polycap™ TC 150, PES, sterile
- Polycap™ TC 150, PES, sterile
- Polycap™ TC 150, PES, sterile
- Polycap™ TC 150, PES, with filling bell, sterile
- Polycap™ TC 150, PES, with filling bell, sterile
- Filtration area:
- 440 cm²
- 440 cm²
- 440 cm²
- 440 cm²
- 440 cm²
- 440 cm²
- 440 cm²
- 930 cm²
- 930 cm²
- 930 cm²
- 930 cm²
- 930 cm²
- 930 cm²
- 1900 cm²
- 1900 cm²
- 1900 cm²
- 1900 cm²
- 1900 cm²
- 1900 cm²
- Max. pressure:
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
Product Family Options
- TypePore sizeConnection
- Polycap™ TC 36, PES, sterile0,2/0,1 µmStepped barb for 6 - 10 mm tubing1 * 1 Items1.010,00 kr
Specifications
Cat. No.515-2531TypePolycap™ TC 36, PES, sterileFiltration area440 cm²Max. pressure4,1 bar - Polycap™ TC 36, PES, sterile0,2/0,2 µmStepped barb for 6 - 10 mm tubing1 * 1 Items1.010,00 kr
Specifications
Cat. No.515-2532TypePolycap™ TC 36, PES, sterileFiltration area440 cm²Max. pressure4,1 bar - Polycap™ TC 36, PES, sterile0,65/0,45 µmStepped barb for 6 - 10 mm tubing1 * 1 Items1.010,00 kr
Specifications
Cat. No.515-2533TypePolycap™ TC 36, PES, sterileFiltration area440 cm²Max. pressure4,1 bar - Polycap™ TC 36, PES, with filling bell, non sterile0,2/0,2 µmMNPT/Stepped barb for 6 - 10 mm tubing1 * 1 Items1.060,00 kr
Specifications
Cat. No.515-2538TypePolycap™ TC 36, PES, with filling bell, non sterileFiltration area440 cm²Max. pressure4,1 bar - Polycap™ TC 36, PES, with filling bell, sterile0,2/0,2 µmStepped barb for 6 - 10 mm tubing1 * 1 Items964,00 kr
Specifications
Cat. No.515-2536TypePolycap™ TC 36, PES, with filling bell, sterileFiltration area440 cm²Max. pressure4,1 bar - Polycap™ TC 36, PES, with filling bell, sterile0,65/0,45 µmStepped barb for 6 - 10 mm tubing1 * 1 Items992,00 kr
Specifications
Cat. No.515-2537TypePolycap™ TC 36, PES, with filling bell, sterileFiltration area440 cm²Max. pressure4,1 bar - Polycap™ TC 36, PES, with filling bell, sterile0,8/0,2 µmStepped barb for 6 - 10 mm tubing1 * 1 Items889,00 kr
Specifications
Cat. No.516-0983TypePolycap™ TC 36, PES, with filling bell, sterileFiltration area440 cm²Max. pressure4,1 bar - Polycap™ TC 75, PES, sterile0,2/0,1 µmStepped barb for 10 - 12 mm tubing1 * 1 Items1.410,00 kr
Specifications
Cat. No.515-2540TypePolycap™ TC 75, PES, sterileFiltration area930 cm²Max. pressure4,1 bar - Polycap™ TC 75, PES, sterile0,2/0,2 µmStepped barb for 6 - 10 mm tubing1 * 1 Items1.410,00 kr
Specifications
Cat. No.515-2541TypePolycap™ TC 75, PES, sterileFiltration area930 cm²Max. pressure4,1 bar - Polycap™ TC 75, PES, sterile0,65/0,45 µmStepped barb for 10 - 12 mm tubing1 * 1 Items1.410,00 kr
Specifications
Cat. No.515-2546TypePolycap™ TC 75, PES, sterileFiltration area930 cm²Max. pressure4,1 bar - Polycap™ TC 75, PES, sterile1,0/1,0 µmStepped barb for 10 - 12 mm tubing1 * 1 Items1.410,00 kr
Specifications
Cat. No.515-2547TypePolycap™ TC 75, PES, sterileFiltration area930 cm²Max. pressure4,1 bar - Polycap™ TC 75, PES, with filling bell, sterile0,2/0,2 µmStepped barb for 6 - 10 mm tubing1 * 1 Items1.470,00 kr
Specifications
Cat. No.515-2544TypePolycap™ TC 75, PES, with filling bell, sterileFiltration area930 cm²Max. pressure4,1 bar - Polycap™ TC 75, PES, with filling bell, sterile0,8/0,2 µmStepped barb for 6 - 10 mm tubing1 * 1 Items964,00 kr
Specifications
Cat. No.516-0984TypePolycap™ TC 75, PES, with filling bell, sterileFiltration area930 cm²Max. pressure4,1 bar - Polycap™ TC 150, PES, sterile0,2/0,1 µmStepped barb for 10 - 12 mm tubing1 * 1 Items1.830,00 kr
Specifications
Cat. No.515-2551TypePolycap™ TC 150, PES, sterileFiltration area1900 cm²Max. pressure4,1 bar - Polycap™ TC 150, PES, sterile0,2/0,2 µmStepped barb for 10 - 12 mm tubing1 * 5 Items7.490,00 kr
Specifications
Cat. No.516-0915TypePolycap™ TC 150, PES, sterileFiltration area1900 cm²Max. pressure4,1 bar - Polycap™ TC 150, PES, sterile0,2/0,2 µmStepped barb for 10 - 12 mm tubing1 * 1 Items1.880,00 kr
Specifications
Cat. No.515-2552TypePolycap™ TC 150, PES, sterileFiltration area1900 cm²Max. pressure4,1 bar - Polycap™ TC 150, PES, sterile0,65/0,45 µmStepped barb for 10 - 12 mm tubing1 * 1 Items1.830,00 kr
Specifications
Cat. No.515-2553TypePolycap™ TC 150, PES, sterileFiltration area1900 cm²Max. pressure4,1 bar - Polycap™ TC 150, PES, with filling bell, sterile0,2/0,2 µmStepped barb for 10 - 12 mm tubing1 * 1 Items1.510,00 kr
Specifications
Cat. No.515-2555TypePolycap™ TC 150, PES, with filling bell, sterileFiltration area1900 cm²Max. pressure4,1 bar - Polycap™ TC 150, PES, with filling bell, sterile0,8/0,2 µmStepped barb for 10 - 12 mm tubing1 * 1 Items1.790,00 kr
Specifications
Cat. No.516-0985TypePolycap™ TC 150, PES, with filling bell, sterileFiltration area1900 cm²Max. pressure4,1 bar