Order Entry
Denmark
ContactUsLinkComponent
 
Disposable weighing dishes
  611-9000
 :  LUDI
undefined
Disposable weighing dishes
  611-9000
 :  LUDI

 

  • Int.Ø×H:
    100×7 mm
  • Pk:
    80

 

 

Aluminium.

  • Diameter: 100 mm
  • Height of rim: 7 mm

For moisture determination.