Order Entry
Denmark
Orders LinkContactUsLinkComponent
Staining jars
Staining jars
Staining jars
Certificates
Staining jars
3 Options Available Below
Product Details & Documents
Soda-lime glass.
  • Three versions
  • For 16 or 10 slides
  • Acc. to Hellendahl or Schiefferdecker
Suitable for standard microscope slides (76×26 mm).
Product Family Options
Product Information
  • Availability
    Price
  • 631-9313
    Staining dishes acc. to Schiefferdecker, for up to 10 slides
    Staining dishes acc. to Schiefferdecker, for up to 10 slides
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 239,00 kr
    239,00 kr
    1 * 1 Items
       239,00 kr
  • 631-9310
    Staining dishes acc. to Hellendahl, for up to 16 slides
    Staining dishes acc. to Hellendahl, for up to 16 slides
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 348,00 kr
    348,00 kr
    1 * 1 Items
       348,00 kr
  • 631-9311
    Staining dishes acc. to Hellendahl, for up to 16 slides, with extension
    Staining dishes acc. to Hellendahl, for up to 16 slides, with extension
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 331,00 kr
    331,00 kr
    1 * 1 Items
       331,00 kr