Order Entry
Denmark
Orders LinkContactUsLinkComponent
Staining dishes
Staining dishes
Certificates
Staining dishes
3 Options Available Below
Product Details & Documents
Soda-lime glass.
  • Removable cover
Staining dish for 76×26 mm slides or slides with width up to 45 mm.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 631-0347
    Staining dish with lid, without tray
    Staining dish with lid, without tray
    90×70×65 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 307,00 kr
    307,00 kr
    1 * 1 Items
       307,00 kr
  • 630-2600
    Staining dish without lid
    Staining dish without lid
    90×70×65 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 177,00 kr
    177,00 kr
    1 * 1 Items
       177,00 kr
  • 631-0346
    Staining dish with lid and tray for 10 slides
    Staining dish with lid and tray for 10 slides
    90×70×65 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 517,00 kr
    517,00 kr
    1 * 1 Items
       517,00 kr
Accessories
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Lid for staining dish
    Lid for staining dish
    631-0864
    - mm
    Log in for availability
    1 * 1 Items - 115,00 kr
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Stainless steel rack for staining jars, for 10 slides
    Stainless steel rack for staining jars, for 10 slides
    631-0348
    - mm
    Log in for availability
    1 * 1 Items - 197,00 kr