Order Entry
Denmark
Orders LinkContactUsLinkComponent
Column end plugs
Column end plugs
Supplier:   VICI JOUR RESEARCH
Column end plugs
Supplier:  VICI JOUR RESEARCH
Certificates
Column end plugs
Column end plugs
3 Options Available Below
Product Details & Documents
These nylon column end plugs fit all columns with 10-32 threads. The long design and threading down to the ferrule ensure safe closing of valuable columns.
  • Available with a 5/16" wrench flat
  • Easy removal
Product Family Options
Product Information
  • Availability
    Price
  • 554-0759
    Column end plug, white
    Column end plug, white
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 155,00 kr
    155,00 kr
    1 * 10 Items
       155,00 kr
  • 554-1146
    Column end plug, black
    Column end plug, black
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 155,00 kr
    155,00 kr
    1 * 10 Items
       155,00 kr
  • 555-3595
    Column end plug, red
    Column end plug, red
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 155,00 kr
    155,00 kr
    1 * 10 Items
       155,00 kr