Flanged fingertight nuts
Fiberglass reinforced PP with PP washers. A number of chromatography systems still require or prefer flanged connections. When the tubing is flanged, the washer never comes in contact with the mobile phase.
- Compatible with all 1/4"-28 flat-bottom ports
- Available for 1/16" and 1/8" tubing
Pressure rating: Up to 30 bar (435 psi).
Pressure rating: Up to 30 bar (435 psi).
Frequently Bought Together