Capillary columns, BP5
Supplier: Trajan Scientific and Medical
Certificates
About this item
A general purpose phase with excellent high temperature characteristics. Popular column used for a wide variety of applications.
5% phenyl/95% dimethyl polysiloxane.
Max. temperature: 340 °C.
Suitable replacement for: CP-Sil 8CB, DB-5, HP-5, MDN-5, PTE-5, Rtx-5, SPB-5, Ultra-2, VB-5, ZB-5.
Frequently Bought Together