Order Entry
Denmark
Orders LinkContactUsLinkComponent
Overalls, Secutex® pro 5/6
Overalls, Secutex® pro 5/6
Overalls, Secutex® pro 5/6
Certificates
Overalls, Secutex® pro 5/6
4 Options Available Below
Product Details & Documents
This particulate resistant disposable overall made from PP-spun fabric and PE laminate protects against fine dusts and splashes of aggressive, liquid chemicals. The overall comes complete with a protective flap over the zip, elasticated hood, back, sleeves and ankles.
  • Generous cut provides excellent freedom of movement, making it extremely comfortable to wear
  • Permeable to air and water vapour
  • Flexible, lint-free, abrasion resistant and anti-static material
Colour: White.
Type 5/6, EN 1149, Cat. III/CE 01
Certifications: CE 0624, Cat. III, Type 5 + 6.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-0657
  • 113-0650
  • 113-0651
  • ZVGA30445-90
  • Anti-static:
  • +
  • +
  • +
  • +
  • Garment application:
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • White
    M
    Log in for availability
    Log in for availability
    1 * 25 Items
    1 * 25 Items - 792,00 kr
    792,00 kr
    1 * 25 Items
       792,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    113-0657
    Anti-static
    +
    Garment application
    Chemical protection|Laboratory|Industrial
  • White
    L
    Log in for availability
    Log in for availability
    1 * 25 Items
    1 * 25 Items - 792,00 kr
    792,00 kr
    1 * 25 Items
       792,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    113-0650
    Anti-static
    +
    Garment application
    Chemical protection|Laboratory|Industrial
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 25 Items
    1 * 25 Items - 792,00 kr
    792,00 kr
    1 * 25 Items
       792,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    113-0651
    Anti-static
    +
    Garment application
    Chemical protection|Laboratory|Industrial
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 25 Items
    1 * 25 Items - 792,00 kr
    792,00 kr
    1 * 25 Items
       792,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    ZVGA30445-90
    Anti-static
    +
    Garment application
    Chemical protection|Laboratory|Industrial