Hoods, Secutex® pro, PP/PE
White
Catalog # 114-3436
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Jump to:
Product Details & Documents
This particulate resistant disposable hood made from PP-spun fabric and PE laminate protects against fine dusts and splashes of aggressive, liquid chemicals.
- Permeable to air and water vapour
- Flexible, lint-free, abrasion resistant and anti-static material
Cat. III, Type 5 + 6
Certifications: Cat. III, Type 5 + 6
Specifications
- Colour:White
- Cat. No.:114-3436
- Garment application:Chemical protection|Laboratory