Order Entry
Denmark
Orders LinkContactUsLinkComponent
Clamps
Supplier:   BOCHEM
Clamps
Supplier:  BOCHEM
Certificates
Clamps
3 Options Available Below
Product Details & Documents
Malleable cast iron, chromed.
  • Jaws have cork inserts to protect clamped items
  • Without bosshead for fixing to stand
  • Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894
Product Family Options
Product Information
  • Spanning width
    Availability
    Price
  • 25 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 146,00 kr
    146,00 kr
    1 * 1 Items
       146,00 kr
  • 40 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 157,00 kr
    157,00 kr
    1 * 1 Items
       157,00 kr
  • 60 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 182,00 kr
    182,00 kr
    1 * 1 Items
       182,00 kr