VWR®, Inoculating Loops and Needles, Disposable
Catalog # 612-9361
Supplier: VWR Collection
Specifications
- Packed:40
- Capacity:1 µl
- Colour:Light green
- Description:Loops, flexible
- Seq:03
- Pk:1.000
Specifications
About this item
PS, sterile.
- Colour coded for easy product identification
- Hexagonal loop shaft improves grip, assisting orientation and making diluting and streaking easier
- Gamma sterilised
The resealable pouch allows loops to be easily accessed any number of times, then re-closed to maintain product integrity. The loops and needles are positioned upside down in the pouch in order to reduce any risk of contamination. Optional pack holder facilitates easy access using one hand, leaving the other hand free to hold a culture plate or tube.
Delivery: Loops are supplied with a calibration certificate in each box.
Packaging: 40 loops/pouch (25 pouches per pack).