Transport carriers, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
These bottle carriers allow for safe transport and storage of equipment, such as bottles, and containers. They include a partition wall and an integral carrying handle.
- Good chemical resistance
- Heat resistant up to +120 °C
- Rigid, with two liquid tight compartments
Colour: white.
Specifications
- Catalog No:
- 139-3110
- 215-8118
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)