Burettes, pellet pattern, lateral and intermediate stopcocks
Supplier: HIRSCHMANN
Certificates
About this item
DURAN®, borosilicate glass, amber, class B.
- Calibrated to deliver 'Ex'
- Short marks at major graduations, white graduations
- Automatic zeroing, lateral stopcock with spindle of PTFE and intermediate stopcock
DIN EN ISO 385
Certifications: DIN EN ISO 385
Delivery: Supplied with 2 litre glass reservoir bottle.
Frequently Bought Together