Wash bottles, narrow neck
Supplier: Kautex
Certificates
About this item
LDPE, translucent with LDPE spray cap.
- Good chemical resistance
- Heat resistant up to 80 °C
- Leakproof
Economical, soft and very easy to press. For use with general solutions such as water, soap, dilute acids and dilute alkalis. The tip can be cut off to increase the flow rate.
Frequently Bought Together