Bottles, wide neck, with standard ground stoppers
Supplier: GLASWARENFABRIK KARL HECHT
Certificates
About this item
Soda-lime glass, clear or brown, with standard ground joint and HDPE stopper.
- Good chemical and physical properties
- For use in applications without high thermal loads
- Replaceable plastic stoppers
Brown coloured to protect light-sensitive materials.
DIN 12039, ISO 4796
Certifications: DIN 12039, ISO 4796
Frequently Bought Together