Ordreindtastning
Denmark
ContactUsLinkComponent
Bottles, wide neck, with screw cap
Bottles, wide neck, with screw cap
Catalog # MURRCO120UC72
Supplier:  MURRAY S
undefined
Bottles, wide neck, with screw cap
Catalog # MURRCO120UC72
Supplier:  MURRAY S

Specifications

  • Udv.Ø:
    62 mm
  • Gevind:
    58/R3
  • Kapacitet:
    120 ml
  • Højde:
    63 mm
  • Pakn:
    72

Specifications

About this item

Clear glass, with black Bakelite closure.

  • Screw closure with paraffin wax lining
  • Good chemical resistance
  • Extra wide opening for easy filling and emptying

Can be used in all applications involving paste-like or powdery materials and liquids.