Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dropping bottles, glass, DURAN®
Dropping bottles, glass, DURAN®
Dropping bottles, glass, DURAN®
Dropping bottles, glass, DURAN®
Soda-lime glass, clear or amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Replaceable NS glass pipette with rubber suction bulb
Amber to protect light-sensitive substances.
Product Family Options
  • Capacity
    Colour
    Height
  • 100 ml
    Clear
    105 mm
    1 * 1 Items
    1 * 1 Items - 121,00 kr
    121,00 kr
    1 * 1 Items
       121,00 kr
  • 100 ml
    Amber
    105 mm
    1 * 1 Items
    1 * 1 Items - 131,00 kr
    131,00 kr
    1 * 1 Items
       131,00 kr
  • 50 ml
    Clear
    79 mm
    1 * 1 Items
    1 * 1 Items - 108,00 kr
    108,00 kr
    1 * 1 Items
       108,00 kr
  • 50 ml
    Amber
    79 mm
    1 * 1 Items
    1 * 1 Items - 113,00 kr
    113,00 kr
    1 * 1 Items
       113,00 kr
Accessories
  •  
    Description
  • SCOT232712405
    Replacement pipette for 100 ml DWK dropping bottle, clear, soda-lime glass, NS 14/15, H 116 mm, Ø 9 mm
    1 * 10 Items - 552,00 kr
  •  
    Description
  • SCOT232711709
    Replacement pipette for 50 ml DWK dropping bottle, clear, soda-lime glass, NS 14/15, H 98 mm, Ø 9 mm
    1 * 10 Items - 530,00 kr