Nalgene™ Carboys, Narrow Neck, Single Use, HDPE
HDPE, translucent with white PP closure.
- Good chemical resistance
- Heat resistant from –100 to +120 °C
- Complies with the requirements of USP class VI, non cytotoxic
Safer than glass. Ideal for storing sterile liquids and pharmaceutical/biotechnology products. The extra wide mouth makes filling and emptying easier.
Eliminate cleaning, prep and set-up costs with ready-to-use Nalgene™ single-use HDPE carboys. Supplied sterile for one-time use, the high-density polyethelyne vessels are ideal for storing sterile fluids and pharmaceutical/biotech products.
HDPE, translucent with white PP closure.
- Good chemical resistance
- Heat resistant from –100 to +120 °C
- Complies with the requirements of USP class VI, non cytotoxic
Safer than glass. Ideal for storing sterile liquids and pharmaceutical/biotechnology products. The extra wide mouth makes filling and emptying easier.
Eliminate cleaning, prep and set-up costs with ready-to-use Nalgene™ single-use HDPE carboys. Supplied sterile for one-time use, the high-density polyethelyne vessels are ideal for storing sterile fluids and pharmaceutical/biotech products.
- Catalog No:
- 215-1182
- NALG342289-0075
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Product Family Options
- DescriptionCapacityTypeMaterialColourExt.ØNeck I-ØHeightThread
- Single-use HDPE carboy, sterile20 LCarboyHDPEWhite284 mm64 mm502 mm83B1 * 6 Items8.590,00 kr
Low Manufacturing Impact Specifications
Cat. No.215-1182Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of Retirement - Single-use HDPE carboy with molded-in handles, sterile33 LSterile CarboyHDPEWhite381 mm64 mm546 mm83B1 * 1 Items2.950,00 kr
Low Manufacturing Impact Specifications
Cat. No.NALG342289-0075Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of Retirement