Carboys, narrow neck, HDPE, with screw cap
: Kautex
HDPE. Natural colour. Blue litre scale.
- Printed litre scale
- Optional stopcock connection
- Suitable for food
- Halal certified
Screwed-on closure. Dust cap serves as a dust protection, but does not seal the carboy. With and without threaded stopcock connection.
Frequently Bought Together