Order Entry
Denmark
Orders LinkContactUsLinkComponent
Double bosshead
Double bosshead
Supplier:   R L ENTERPRISES
Double bosshead
Supplier:  R L ENTERPRISES
Certificates
Double bosshead
2 Options Available Below
Product Details & Documents
Bossheads made from nickel plated cast alloy.
  • Either with or without blue plastic headed nickel plated screws
  • For use with rods up to 16 mm
Product Family Options
Product Information
  • Material
    For rod Ø
    Availability
    Price
  • 241-7867
    Bosshead with blue plastic headed screw
    Bosshead with blue plastic headed screw
    Cast alloy
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 244,00 kr
    244,00 kr
    1 * 1 Items
       244,00 kr
  • RLAA2005/1PL
    Bosshead without blue plastic headed screw
    Bosshead without blue plastic headed screw
    Cast alloy
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 197,00 kr
    197,00 kr
    1 * 1 Items
       197,00 kr