Beakers, Philips, DURAN®
Catalog # 213-0214
Supplier: DWK Life Sciences
Supplier #: 211413609
Log in for availability
About this item
DURAN®, borosilicate glass 3.3, clear, with spout.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Thick-walled, with printed graduations and labelling area.
Frequently Bought Together