Order Entry
Denmark
ContactUsLinkComponent
Rechargeable Batteries for Cordless/Mobile Telephones
Rechargeable Batteries for Cordless/Mobile Telephones
Catalog # 139-2271
undefined
Rechargeable Batteries for Cordless/Mobile Telephones
Catalog # 139-2271
Specifications
  • Type:
    Mignon AA/HR6
  • Voltage:
    1,2 V
  • Capacity:
    1700 mAh
  • Description:
    Rechargable NiMH battery for T399 phone
  • Environmentally Preferable:
  • Pk:
    2
Specifications
About this item
Nickel Hydride (NiMH), AA/HR6 1,2 V. These batteries are compatible with a wide range of cordless phones including Alcatel, Ascom, Audioline, Binatone, Bosch, BT, Deutsche Telekom, Hagenuk, Olympia, Panasonic, Philips, Samsung, Sanyo, Siemens, Swatch.