Size exclusion chromatography columns, GPC LF series
: Resonac
Gel permeation columns (GPC) packed with tetrahydrofuran (THF), which has a wide pore size distribution designed for high linearity of the calibration curve.
- Enables analysis over a broad range of molecular weights (100 to 2 million)
- Highly linear calibration curve without inflection points
- Molecular weight distribution can be determined with precision
- Catalog No:
- 554-1769
- 554-1764
- 554-1770
- 554-1766
- Description:
- LF-604
- LF-804
- LF-404
- LF-G
- Max. pressure:
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- Temperature limit:
- 20 - 60 °C
- 20 - 60 °C
- 20 - 60 °C
- 20 - 60 °C
- USP code:
- L21
- L21
- L21
- L21
Frequently Bought Together