Order Entry
Denmark
Orders LinkContactUsLinkComponent
Murexide

Murexide

Glass bottle for solids, 5 g
Catalog # ROTHT124.1
Marketsource
Supplier:   Roth Carl
Murexide
Catalog # ROTHT124.1
Supplier:  Roth Carl
Marketsource
CAS Number:  3051-09-0
Murexide

Glass bottle for solids, 5 g

CAS Number:  3051-09-0
Price
250,17 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
  • CAS Number:3051-09-0
  • Formula:C₈H₈N₆O₆
  • MDL Number:MFCD00012777
  • Molecular Weight:284.19 g/mol
  • Merck Index:13,06330
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:221-266-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.