Order Entry
Denmark
Orders LinkContactUsLinkComponent
 
Murexide
Certificates
Murexide

3 Options Available Below

Product Details & Documents

CAS No.: 3051-09-0 Molecular Formula: C8H8N6O6 Formula Weight: 284.19 Melting Point: 300° Beilstein Reference: 3582175 Merck Reference: 14,6306 MDL No.: MFCD00012777

Synonyms:  Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
  • CAS Number:3051-09-0
  • Formula:C₈H₈N₆O₆
  • MDL Number:MFCD00012777
  • Molecular Weight:284.19 g/mol
  • Merck Index:14,6306
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:221-266-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 612,00 kr
    612,00 kr
    1 * 10 g
       612,00 kr

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 2.030,00 kr
    2.030,00 kr
    1 * 50 g
       2.030,00 kr

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 7.650,00 kr
    7.650,00 kr
    1 * 250 g
       7.650,00 kr