Order Entry
Denmark
Orders LinkContactUsLinkComponent
 
Murexide ACS
Certificates
Murexide ACS

4 Options Available Below

Product Details & Documents
Synonyms:  Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
  • CAS Number:3051-09-0
  • Formula:C₈H₈N₆O₆
  • MDL Number:MFCD00012777
  • Molecular Weight:284.19 g/mol
  • Merck Index:13,06330
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:221-266-6
Specifications for Product Family

Specification Test Results

  • Appearance:
    Bordeaux to red-purple Powder
  • Infrared spectrum:
    Conforms
  • Suitability:
    (as complexom. indicator) Passes test
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.