Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Thymolphthalein, pure indicator

Thymolphthalein, pure indicator
Certificates
Thymolphthalein, pure indicator

3 Options Available Below

Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.54 g/mol
  • Merck Index:12,09542
  • Melting Pt:251…253 °C
  • Storage Temperature:Ambient
  • EINECS:204-729-7

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 %
  • Sulfated ash:
    ≤0.5 %
  • Visual transition interval:
    from pH 8.8 (colorless) to pH 10.5 (blue)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 489,00 kr
    489,00 kr
    1 * 10 g
       489,00 kr

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 1.680,00 kr
    1.680,00 kr
    1 * 50 g
       1.680,00 kr

  • Glass bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 4.840,00 kr
    4.840,00 kr
    1 * 250 g
       4.840,00 kr