Methyl cellulose, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9004-67-5
:
MDL Number:MFCD00081763
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Viscosity (2% aqueous solution at 25 °C):~15 centipoises
- Loss on Drying:≤5,0%
- Methoxy groups:27,5 - 31,5%
Specification Test Results
Frequently Bought Together