Order Entry
Denmark
ContactUsLinkComponent
α-Hydroxy-o-tolyl-β-D-glucopyranoside, powder
α-Hydroxy-o-tolyl-β-D-glucopyranoside, powder
Supplier:  MP Biomedicals
Certificates
α-Hydroxy-o-tolyl-β-D-glucopyranoside, powder
Supplier:  MP Biomedicals
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Identity Test :
    Passes
  • FTIR:
    Conforms to Standard
  • 194 - 204 °C:
    Melting Point
  • Optical Rotation:
    −65° ± 5° (c=3, water)