α-Hydroxy-o-tolyl-β-D-glucopyranoside
Catalog # SERA34600.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, D(-)-Salicin, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol, D(−)-Salicin
:
:286.28 g/mol
EINECS:205-331-6
CAS Number:138-52-3
Formula:C₁₃H₁₈O₇
MDL Number:MFCD00006590
Melting Pt:199…201 °C
Density:1.42 g/cm³ (20 °C)
Merck Index:12,08476