Order Entry
Denmark
ContactUsLinkComponent
 
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid), Millipore®
Catalog # 324626-25
CAS Number:  
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid), Millipore®
Catalog # 324626-25
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Merck Index:13,03560
Melting Pt:240…244 °C
 :380.35 g/mol
Storage Temperature:Ambient
EINECS:200-651-2
CAS Number:67-42-5
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291