Order Entry
Denmark
ContactUsLinkComponent
Human/Mouse/Rat Recombinant BMP-2 (from E. coli)
Human/Mouse/Rat Recombinant BMP-2 (from E. coli)
Certificates
Human/Mouse/Rat Recombinant BMP-2 (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Rat
    Mouse
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity Assay: Alkaline phosphatase activity induced in ATDC-5 cells, ED₅₀ ≤250 ng/ml, Bioactivity ≥4,0×10³ units/mg
  • Gene ID:
    P12643 / P21274 / P49001
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    BMP-2A
  • Protein/peptide name:
    H/M/R BMP-2
  • Purity:
    ≥95%
  • Molecular weight:
    13,0/26,1 kDa
  • Sequence:
    MQAKHKQRKR LKSSCKRHPL YVDFSDVGWN DWIVAPPGYH AFYCHGECPF PLADHLNSTN HAIVQTLVNS VNSKIPKACC VPTELSAISM LYLDENEKVV LKNYQDMVVE GCGCR
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together