Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant IL-3 (CTGrade GMP) (from E. coli)
Human Recombinant IL-3 (CTGrade GMP) (from E. coli)
New Product
Certificates
Human Recombinant IL-3 (CTGrade GMP) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,05
  • Biological activity:
    Bioactivity assay: Proliferation of TF-1, ED50 ≤2 ng/ml, Bioactivity ≥4,2×10³ IU/µg
  • Gene ID:
    P08700
  • Amino acid number:
    134
  • Reconstitution instructions:
    Sterile water at 0.1 - 0.5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    MCGF|P-cell stimulating factor|HCGF|Multi-CSF
  • Protein/peptide name:
    IL-3
  • Purity:
    ≥97%
  • Molecular weight:
    15,2 kDa
  • Sequence:
    MAPMTQTTPL KTSWVNCSNM IDEIITHLKQ PPLPLLDFNN LNGEDQDILM ENNLRRPNLE AFNRAVKSLQ NASAIESILK NLLPCLPLAT AAPTRHPIHI KDGDWNEFRR KLTFYLKTLE NAQAQQTTLS LAIF
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 150 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    CTGrade GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together