Millex® Syringe Filters, MCE Membrane
Supplier: Merck
Certificates
About this item
Filters for sterilising or clarifying biological solutions. The larger filtration area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.
Manufacturing occurs in a controlled environment using an automated process. Provided with a certificate of quality.
Specifications
- Catalog No:
- MILFSLHA033SS
- MILFSLHA033SB
- MILFSLGSV255F
- MILFSLGS033SS
- MILFSLHAM33SS
- MILFSLGS033SB
- MILFSLAAV255F
- MILFSLGSM33SS
- MILFSLGL0250S
- MILFSLAA033SB
- MILFSLAA033SS
- Max. operating pressure:
- 10 bar
- 10 bar
- 10 bar
- 10 bar
- 10 bar
- 10 bar
- 3 bar
- 10 bar
- 5 bar
- 10 bar
- 10 bar
- Max. temperature:
- 45 °C
- 45 °C
- 45 °C
- 45 °C
- 45 °C
- 45 °C
- 45 °C
- 45 °C
- 45 °C
- 45 °C
- 45 °C