Order Entry
Denmark
ContactUsLinkComponent
Millicell® µ-Migration Assay Kit
Millicell® µ-Migration Assay Kit
Catalog # MILFMMA205
Supplier:  Merck
CAS Number:  
undefined
Millicell® µ-Migration Assay Kit
Catalog # MILFMMA205
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Millicell® µ-migration assay kit
  • Species reactivity:
    Rat, mouse, human
  • Shipping conditions:
    Dry Ice

Specifications

About this item

This Kit overcomes the limitations of traditional multiwell migration assays. Its microfluidic, low-volume technology promotes a stable, diffusion-generated concentration gradient that is consistently linear and lasts for more than 48 hours.