Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microplates
Cell Culture Plates
Plexiglas® Rings
Plexiglas® Rings
Plexiglas® Rings
Catalog # MILFPR0001400
Supplier:
Merck
Expand image
Plexiglas® Rings
Catalog # MILFPR0001400
Supplier:
Merck
Specifications
Description:
Rings without hole, Ø 14 mm, 80 μm pore size
Seq:
01
Pk:
50
Specifications
About this item
Product Details
Product Details