Order Entry
Denmark
ContactUsLinkComponent
Light Duty Measuring Jugs
Light Duty Measuring Jugs
Catalog # COLDMEA4002G
Supplier #: MEA4002G
undefined
Light Duty Measuring Jugs
Catalog # COLDMEA4002G
Supplier #: MEA4002G
Log in for availability
Select an Option
170 mm
Select an Option
2 L
Select an Option
Light duty measuring jug, stainless steel 304
Select an Option
200 mm
Select an Option
03
Select an Option
1
Select an Option
320 g
About this item
Stainless steel measuring jugs meet international pharma quality compliance regulations.
  • All sizes available as standard, off-the-shelf for delivery within days
  • 304 pharmaceutical grade stainless steel
  • Graduated in pints and litres
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Material Grade, Surface Finish, and TSE certification available upon request
Made from 304 grade stainless steel, these measuring jugs are tapered and come with either an open or closed handle. They feature crevice-free and pit-free interiors and are bright polished throughout.
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information. Measuring jugs are available in Heavy Duty, Light Duty, and Light Duty with rimmed foot formats. The option of rimmed feet is available with the closed handle version from 0.5 litre upwards.
Frequently Bought Together