Order Entry
Denmark
Orders LinkContactUsLinkComponent
Antibiotic Sensitivity Discs
 :   HIMEDIA
Antibiotic Sensitivity Discs
 :  HIMEDIA
Antibiotic Sensitivity Discs
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
Product Family Options
  • Description
    Pk
  • Ampicillin, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Azithromycin, 15 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Azlocillin, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 172,00 kr
    172,00 kr
    1 * 50 Items
       172,00 kr
  • Aztreonam, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Cefalexin (Cephalexin), 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Cefazolin, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Cefixime, 5 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Cefotaxime, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Cefoxitin, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Cefpodoxime, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Cefradine, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Ceftizoxime, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Chloramphenicol, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Ciprofloxacin, 5 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Clarithromycin, 15 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Clotrimazole, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Co-Trimoxazole (Sulpha/Trimethoprim) 25 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Colistin (methane sulphonate), 25 mcg
    50
    1 * 50 Items
    1 * 50 Items - 179,00 kr
    179,00 kr
    1 * 50 Items
       179,00 kr
  • Doripenem, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 1.050,00 kr
    1.050,00 kr
    1 * 50 Items
       1.050,00 kr
  • Doxycycline hydrochloride, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Fluconazole, 25 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Fosfomycin, 50 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Gatifloxacin, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Gentamicin, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Ketoconazole, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Levofloxacin, 5 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Metronidazole, 5 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Miconazole, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Moxifloxacin, 5 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Nitrofurazone, 100 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Norfloxacin, 10 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Nystatin, 100 units
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Ofloxacin, 5 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr
  • Tobramycin, 30 mcg
    50
    1 * 50 Items
    1 * 50 Items - 145,00 kr
    145,00 kr
    1 * 50 Items
       145,00 kr