Order Entry
Denmark
Orders LinkContactUsLinkComponent
Starch (from arrowroot), powder

Starch (from arrowroot), powder

Poly bottle, 0,5 kg
Catalog # SPCMS1551-500GM
Supplier:   Spectrum Chemical
Starch (from arrowroot), powder
Catalog # SPCMS1551-500GM
Supplier:  Spectrum Chemical
CAS Number:  9005-25-8
Starch (from arrowroot), powder

Poly bottle, 0,5 kg

CAS Number:  9005-25-8
Price
997,00 kr
Log in for availability
1 * 0,5 kg
Product Details & Documents

Starch, Arrowroot, Powder is used in talcum powders and hair dyes. It is added to moisturizers as a thickening agent and to help active ingredients penetrate the upper levels of the skin. This product is used in cosmetics to help moisturizers penetrate the skin. It appears as a fine, white powder. It may have other uses similar to cornstarch. In cosmetics, it is used as an abrasive.

  • CAS Number:9005-25-8
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    0,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.