
Palladium(II) chloride 0,4% in hydrochloric acid 13.5% Reag. Ph. Eur. 1061501
Catalog # 87884.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
ADR:8,II
:177.33 g/mol
EINECS:231-596-2
UN:3264
:GHS07
Storage Temperature:Ambient
CAS Number:7647-10-1
Formula:PdCl₂
MDL Number:MFCD00003558