Sodium azide 1% in aqueous solution
Catalog # 786-299
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: Sodium triazide
:
:65.01 g/mol
EINECS:247-852-1
:GHS07
CAS Number:26628-22-8
Formula:NaN₃
MDL Number:MFCD00003536